Anti-CD74 Mouse Monoclonal Antibody [clone: PIN1]

Proveedor: Cayman Chemical
10011432-100 10011432-25
CAYM10011432-100EA 746 EUR
CAYM10011432-100 CAYM10011432-25
Anti-CD74 Mouse Monoclonal Antibody [clone: PIN1]
Anticuerpos
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: PIN1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR