Anti-CaMKII Mouse Monoclonal Antibody [clone: 6G9]

Proveedor: Cayman Chemical
10011437-100 10011437-25
CAYM10011437-100EA 752 EUR
CAYM10011437-100 CAYM10011437-25
Anti-CaMKII Mouse Monoclonal Antibody [clone: 6G9]
Anticuerpos
Type: Primary
Antigen: CaMKII
Clonality: Monoclonal
Clone: 6G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR