Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1D12]

Proveedor: Cayman Chemical
170101-100 170101-500
CAYM170101-100EA 546 EUR
CAYM170101-100 CAYM170101-500
Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1D12]
Anticuerpos
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-1D12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: brown bullhead
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR