Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1H2]

Proveedor: Cayman Chemical
170265-100 170265-500
CAYM170265-100EA 546 EUR
CAYM170265-100 CAYM170265-500
Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1H2]
Anticuerpos
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-1H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Gulf Sturgeon, Lake Sturgeon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR