Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: N/A]

Proveedor: Cayman Chemical
189542-200 189542-50
CAYM189542-200EA 532 EUR
CAYM189542-200 CAYM189542-50
Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: N/A]
Anticuerpos
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: N/A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b 
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR