Anti-CD154 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MR1]

Proveedor: Tonbo Biosciences
50-1541-U025 50-1541-U100
50-1541-U025EA 41.6 EUR
50-1541-U025 50-1541-U100
Anti-CD154 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MR1]
Anticuerpos
The MR1 antibody is specific for mouse CD154, a 39 kD glycoprotein that is primarily expressed on activated T cells. CD154 (CD40 ligand, gp39) binds to CD40 expressed on antigen presenting cells and mediates important costimulatory functions between these cells. Signals generated via cognate interactions between CD154-CD40 on activated T and B cells are required for T-dependent B cell responses. The MR1 antibody is reported to block interaction of CD154 with CD40.

Type: Primary
Antigen: CD154
Clonality: Monoclonal
Clone: MR1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR