Gas detection tubes for carbon disulphide
Proveedor: Dräger
8101891 6728351 CH23201
151-0183EA 92.2 EUR
151-0183 DRAE6728351 118-0339
Gas detection tubes for carbon disulphide
Tubos per gas
Operating conditions: 0 to 40 °C ; up to 30 mg/l H₂O and for tube 5/a: 3 to 15 mg/l H₂O absolute humidity.
  • Data below representative for use with an Accuro® pump
  • Cross sensitivity for tube 3/a hydrogen sulphide in the TLV range retained in the pre-layer (does not interfere)
  • For tubes carbon disulphide 5/a and 30/a it is impossible to measure carbon disulphide in the presence of hydrogen sulphide
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Order Now
ESPECIFICACIONES
Modelo Carbon disiphide 3/a Carbon disiphide 5/a Carbon disiphide 30/a
Environmentally Preferable
Cambio de color Pale blue to yellow green White to brown green Pale blue to brown
Tiempo de medición 2 minutes 3 minutes 1 minute
Nr. de golpes 15 - 1 11 6
Desviación Estándar ±30% ±10 - 15 % ±15 - 20 %
Rango estándar 3 - 95 ppm 5 - 60 ppm 0,1 - 10 mg/l