Gas detection tubes for carbon monoxide
Proveedor: Dräger
6733051 CH20601 CH25601 CH29901 CH19701
151-0162EA 82.2 EUR
151-0162 151-0190 151-3159 118-0347 151-0188
Gas detection tubes for carbon monoxide
Tubos per gas
To measure the 'spot concentrations' of carbon monoxide for use with an Accuro pump. Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Operating conditions: 0 to 50 °C ; up to 50 mg/L H₂O absolute humidity ; tube 2/a: 0 to 50 °C ; 2 to 20 mg/L H₂O absolute humidity.
Order Now
ESPECIFICACIONES
Modelo Carbon monoxide 2/a Carbon monoxide 10/b Carbon monoxide 5/c Carbon monoxide 0.3%/b Carbon monoxide 8/a
Environmentally Preferable
Cambio de color White to brownish White to brownish green White to brownish green White to brownish green White to pale brown
Tiempo de medición 4 minutes/50 seconds 20 seconds/4 minutes 50 seconds/2,5 minutes 30 seconds 2 minutes
Nr. de golpes 10/2 1/10 1/5 1 10
Desviación Estándar ±10 - 15 % ±10 - 15 % ±10 - 15 % ±10 - 15 % ±10 - 15 %
Rango estándar 2 - 60/25 - 300 ppm 100 - 3000/10 - 300 ppm 100 - 700/5 - 150 ppm 0,3 - 7 vol %   8 - 150 ppm