Anti-CD20 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H7]

Proveedor: Tonbo Biosciences
50-0209-T025 50-0209-T100 50-0209-T500
50-0209-T025EA 63.9 EUR
50-0209-T025 50-0209-T100 50-0209-T500
Anti-CD20 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2H7]
Anticuerpos
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR