Anti-CD5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: L17F12]

Proveedor: Tonbo Biosciences
67-0058-T025 67-0058-T100 67-0058-T500
67-0058-T025EA 137 EUR
67-0058-T025 67-0058-T100 67-0058-T500
Anti-CD5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: L17F12]
Anticuerpos
The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno CD5
Environmentally Preferable
Nombre del antígeno CD5 molecule
Sinónimos de antígenos CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado PerCP (Peridinin-Chlorophyll Protein Complex)
Clon L17F12
Reactividad Human
Huésped Mouse
ID del gen 921
Isotipo IgG2a kappa
Citometría de flujo Yes
Adsorción cruzada No
Temperatura de almacenamiento Store undiluted at 2-8 °C, protect from light, do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR