Anti-CD25 Rat Monoclonal Antibody (Biotin) [clone: PC61.5]
Proveedor: Tonbo Biosciences
30-0251-U100 30-0251-U500 30-0251-U025
30-0251-U100EA 81 EUR
30-0251-U100 30-0251-U500 30-0251-U025
Anti-CD25 Rat Monoclonal Antibody (Biotin) [clone: PC61.5]
Anticuerpos
The PC61.5 antibody is specific for mouse CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Expression of CD25 varies during developmental stages of T and B cells, is induced on activated mature T and B cells, and is present on subsets of dendritic cells. CD25 signaling as partMore Product Information