Dual Vespel® ring inlet seals
Proveedor: Restek
21242 21238 21239 21240 21241 21243 21244 21245 21246 21247 21248 21249
REST21242EA 94 EUR
REST21242 REST21238 REST21239 REST21240 REST21241 REST21243 REST21244 REST21245 REST21246 REST21247 REST21248 REST21249
Dual Vespel® ring inlet seals
Juntas Sellos mecánicos
A ring inlet seal greatly improves injection port performance as it stays sealed even after repeated temperature cycles, without retightening the reducing nut. This seal features two soft Vespel® rings, one embedded in its top surface and the other embedded in its bottom surface. These rings eliminate the need for a washer and ensure very little torque is needed to make a leak-tight seal.