Burners, Teclu
Proveedor: JUCHHEIM KARL KURT
VL01262DE VL01262DP
JUCHVL01262DEEA 0 EUR
JUCHVL01262DE JUCHVL01262DP
Burners, Teclu
Calentadores Mecheros de gas
Teclu burners with needle valve.
Height: 150 mm
Weight: 325 g