Laboratory washer-disinfector, G 7823
Proveedor: MIELE
6174430 6174420
141-0498EA 0 EUR
141-0498 141-0497
Laboratory washer-disinfector, G 7823
Lavadoras de vidrio
The G 7823 serie is designed specially to meet the needs of small to medium sized hospitals. With a width of only 900 mm, these machines are the perfect professional solution for daily instrument preparation in situations where space is limited. For central instrument processing, one or more Miele washer-disinfectors can be installed, depending on the size of the hospital. Placing the machines sideby-side can increase the output capacity and optimise flexibility.
  • Hygienic freshwater system with water change after each cleaning phase
  • Cleaning, disinfection and drying in a closed system
  • High-capacity water softener (external)
  • Integrated 6-pin printer to record key process parameters
  • Flow meter counter for control of water intake
  • Spray arm monitoring
  • Direct docking of mobile units to water circuit
  • Electrical door lock
  • Heater elements outside wash chamber
  • Low heat and sound emissions thanks to double insulation
  • 2 powerful circulation pumps
  • Triple filtration with large surface filter, coarse filter and micro-fine filter
  • Class H 13 high temperature HEPA filter, located upstream from the cabinet, prevents the unwanted air-borne particles from room air
  • Serial interfaces RS 232 for process documentation, optical interface for customer service and servicing work
The single chamber principle of all Miele washer-disinfectors (each machine cleans, disinfects and dries independently of the others) makes them particularly versatile and economical. Every instrument is thoroughly cleaned and reliably disinfected.
Construction: Freestanding appliance with single drop down door,the frame is in AISI 304 stainless steel, external casing in AISI 304 stainless steel, grain brushed to 220 grade.
Certificaciones: Comprehensive safety features in accordance with EN ISO 15883
Información para pedidos: Baskets and inserts must be ordered separately.