Safety shoes, lace-up, SK Grip, Nero Grip
Proveedor: U-Power
SG20154/38 SG20154/39 SG20154/40 SG20154/41 SG20154/42 SG20154/43 SG20154/44 SG20154/45 SG20154/46 SG20154/47
UPOWSG20154/38EA 83.5 EUR
UPOWSG20154/38 UPOWSG20154/39 UPOWSG20154/40 UPOWSG20154/41 UPOWSG20154/42 UPOWSG20154/43 UPOWSG20154/44 UPOWSG20154/45 UPOWSG20154/46 UPOWSG20154/47
Safety shoes, lace-up, SK Grip, Nero Grip
Zapatos y botas
Safety shoes made from full grain water resistant leather and a lightweight, extremely flexible, slip resistant PU/PU U-Grip sole.
  • Anatomically shaped Polysoft insole made from soft PU, with pockets in the foot support area
  • PU/PU walking sole with 90° angle blocks and multilevel stabilising channels to for extremely high anti-slip features; TPU anti twist insert for additional support
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective composite toecap (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificaciones: EN 20345:2011; S3, SRC