Rutenio 5% on alumina, en polvo

Proveedor: Thermo Scientific

11749.06 11749.14 11749.22
11749.06EA 70 EUR
11749.06 11749.14 11749.22
Rutenio 5% on alumina, en polvo
Rutenio

CAS No.: 7440-18-8 Physical Form: Powder Appearance: Greyish white Odour: Odourless MDL No.: MFCD00011207


Applications: Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules

Formula: Ru
MW: 101,07 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011207
Núm. CAS: 7440-18-8
EINECS: 231-127-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR