Ruthenium(III) nitrosylnitrate (≥31,3% Ru)
Proveedor: Thermo Fisher Scientific
|
Peligro
|
12175.03
12175.06
12175.14
12175.03EA
143
EUR
12175.03
12175.06
12175.14
Ruthenium(III) nitrosylnitrate (≥31,3% Ru)
Ruthenium(III) nitrosylnitrate
CAS No.: 34513-98-9
Molecular Formula: N4O10Ru
Formula Weight: 317.09
Physical Form: Crystalline
Appearance: Red-brown
Odour: Odourless
Melting Point: dec.
UN No.: UN1477
MDL No.: MFCD00016313
Formula:
N₄O₁₀Ru MW: 317.09 g/mol Pto. de fusión: Dec. Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00016313 Núm. CAS: 34513-98-9 EINECS: 252-068-8 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...