Cerium(III) acetylacetonate hydrate

Proveedor: Thermo Fisher Scientific
Atención

21116.22 21116.36
21116.22EA 295 EUR
21116.22 21116.36
Cerium(III) acetylacetonate hydrate
Cerium(III) acetylacetonate hydrate

CAS No.: 206996-61-4 Molecular Formula: C15H21CeO6.xH2O Formula Weight: 437.44(anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: Yellow to red Melting Point: 131-132° MDL No.: MFCD00150164

Formula: C₁₅H₂₁CeO₆·xH₂O
Pto. de fusión: 131…132 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00150164
Núm. CAS: 206996-61-4
EINECS: 239-720-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR