Mullite

Proveedor: Thermo Fisher Scientific

22881.22 22881.36
22881.22EA 67.1 EUR
22881.22 22881.36
Mullite
Mullite

CAS No.: 1302-93-8 Molecular Formula: 3Al2O3.2SiO2 (formula approx.) Physical Form: -325 Mesh Powder MDL No.: MFCD00058866

Formula: ∼3Al₂O₃·2SiO₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00058866
Núm. CAS: 1302-93-8
EINECS: 215-113-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR