Nickel tin oxide dihydrate

Proveedor: Thermo Scientific
Peligro

39452.22 39452.36
39452.22EA 110 EUR
39452.22 39452.36
Nickel tin oxide dihydrate
Nickel tin oxide dihydrate

CAS No.: 12035-38-0 Molecular Formula: NiSnO3.2H2O Formula Weight: 261.43 (225.40anhy) Physical Form: Powder Appearance: Pale green Odour: Odourless MDL No.: MFCD00054011

Formula: NiSnO₃·2H₂O
MW: 261,43 g/mol (225,40 anhy)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00054011
Núm. CAS: 12035-38-0
EINECS: 234-824-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR