Aluminum acrylate
Proveedor: Thermo Fisher Scientific
|
Atención
|
42003.06
42003.14
42003.22
42003.06EA
57.6
EUR
42003.06
42003.14
42003.22
Aluminum acrylate
Aluminum acrylate
CAS No.: 15743-20-1
Molecular Formula: Al(H2C=CHCOO)3
Formula Weight: 243.17
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
MDL No.: MFCD00078421
Formula:
Al(H₂C=CHCOO)₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00078421 Núm. CAS: 15743-20-1 EINECS: 239-836-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...