Zirconium carbonate basic hydrate ≥97%
Proveedor: Thermo Scientific
43245.36
43245.A3
43245.36EA
71.5
EUR
43245.36
43245.A3
Zirconium carbonate basic hydrate ≥97%
Zirconium carbonate basic hydrate
CAS No.: 57219-64-4
Molecular Formula: [ZrO2]2.CO2.xH2O
Formula Weight: 290.45(anhy)
Physical Form: Powder
Density: 2.2
Appearance: White
MDL No.: MFCD00172265
Formula:
C₄H₄O₁₂Zr MW: 335,288 g/mol Densidad: 2,2 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00172265 Núm. CAS: 57219-64-4 EINECS: 260-633-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...