Silver Protein strong, en polvo para histología
Proveedor: Thermo Scientific
Sinónimos:
Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver
Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.
- Silver content: min 7.5%
- Hygroscopic
- Light Sensitive
- Store under Nitrogen
- Soluble in water. Slightly soluble in methanol, diethyl ether
CAS No.: 9008-42-8
Physical Form: Powder
Sensitivity: Light Sensitive; Hygroscopic
Appearance: Pale yellow to brown
MDL No.: MFCD00132268
Applications: Histological stain for microscopic analysis
Formula:
Ag (with protein) MW: 107,87 g/mol |
Número MDL:
MFCD00132268 Núm. CAS: 9008-42-8 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...