Hydridotetrakis(triphenylphosphine)rhodium(I) (≥8,9% Rh)
Proveedor: Thermo Fisher Scientific
|
Atención
|
44030.03
44030.06
44030.03EA
404
EUR
44030.03
44030.06
Hydridotetrakis(triphenylphosphine)rhodium(I) (≥8,9% Rh)
Hydridotetrakis(triphenylphosphine)rhodium(I)
CAS No.: 18284-36-1
Molecular Formula: RhH[P(C6H5)3]4
Formula Weight: 1153.09
Physical Form: Powder
Sensitivity: Air Sensitive
Melting Point: 145-147° dec.
Solubility: Soluble in chloroform, toluene
MDL No.: MFCD00015867
Formula:
C₇₂H₆₁P₄Rh Pto. de fusión: 145…147 °C (desc.) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00015867 Núm. CAS: 18284-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...