Aluminum oxalate hydrate ≥99%
Proveedor: Thermo Scientific
|
Atención
|
44149.30
44149.09
44149.18
44149.A1
44149.30EA
414
EUR
44149.30
44149.09
44149.18
44149.A1
Aluminum oxalate hydrate ≥99%
Aluminum oxalate hydrate
Purity: 99%
CAS No.: 814-87-9
Molecular Formula: C6Al2O12.xH2O
Formula Weight: 318.02(anhy)
Physical Form: Powder/chunks
Appearance: White
MDL No.: MFCD00050791
Formula:
C₆Al₂O₁₂·xH₂O MW: 318,017 g/mol (anhy) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00050791 Núm. CAS: 814-87-9 EINECS: 212-407-2 Índice Merck: 14,355 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...