Calcium-α-D-isosaccharinate ≥98%
Proveedor: Thermo Fisher Scientific
44371.06
44371.14
44371.06EA
473
EUR
44371.06
44371.14
Calcium-α-D-isosaccharinate ≥98%
Calcium-alpha-D-isosaccharinate
Purity: 98%
Molecular Formula: C12H22CaO12
Formula Weight: 398.38
Physical Form: Crystalline
Appearance: Pale brown
MDL No.: MFCD04039767
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...