4-Methylbenzhydrylamine hydrochloride, polymer supported 0,5 - 1,0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Proveedor: Thermo Scientific
|
Atención
|
44455.06
44455.14
44455.06EA
86.3
EUR
44455.06
44455.14
4-Methylbenzhydrylamine hydrochloride, polymer supported 0,5 - 1,0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
4-Methylbenzhydrylamine hydrochloride, polymer supported
Physical Form: Beads
Appearance: Off-white to pale yellow
MDL No.: MFCD00801150
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...