Titanium(IV) 2-ethylhexanoate ≥97%
Proveedor: Thermo Scientific
|
Atención
|
44602.22
44602.36
44602.XK
44602.22EA
407
EUR
44602.22
44602.36
44602.XK
Titanium(IV) 2-ethylhexanoate ≥97%
Titanium(IV) 2-ethylhexanoate
Purity: 97%
CAS No.: 3645-34-9
Molecular Formula: C32H60O8Ti
Formula Weight: 623.75
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD05663781
Formula:
C₃₂H₆₀O₈Ti Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD05663781 Núm. CAS: 3645-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...