Phenyl phosphate disodium salt ≥98%
Proveedor: Thermo Scientific
Sinónimos:
Phosphoric acid phenyl ester disodium salt, Phosphoric acid monophenyl ester disodium salt
Purity: 98%
CAS No.: 3279-54-7
Molecular Formula: C6H5Na2O4P.xH2O
Formula Weight: 218.06(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Pale yellow
Beilstein Reference: 4167154
MDL No.: MFCD00002133
Applications: Used in phosphatase determinations
Formula:
C₆H₅Na₂O₄P·xH₂O MW: 218,06 g/mol (anhy) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00002133 Núm. CAS: 3279-54-7 EINECS: 221-917-4 Índice Merck: 14,3357 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...