Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform (20,6% Pd)
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct, Tris(dibenylideneacetone)dipalladium-chloroform, Tris(dibenzylideneacetone)dipalladium-chloroform adduct
CAS No.: 52522-40-4
Molecular Formula: C51H42O3Pd2.CHCl3
Formula Weight: 1035.11
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Melting Point: 131-135°
MDL No.: MFCD00075479
Formula:
C₅₁H₄₂O₃Pd₂·CHCl₃ MW: 1035,11 g/mol Pto. de fusión: 131…135 °C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00075479 Núm. CAS: 52522-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...