Polyethyleneimine, M.W. 1,200 ≥99%, branched
Proveedor: Thermo Fisher Scientific
|
Atención
|
Sinónimos:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
45594.14
45594.22
45594.36
45594.14EA
45.5
EUR
45594.14
45594.22
45594.36
Polyethyleneimine, M.W. 1,200 ≥99%, branched
Polyethyleneimine
Purity: 99%
CAS No.: 9002-98-6
Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y
Physical Form: Liquid
Appearance: Pale yellow
Odour: Amine-like
MDL No.: MFCD00803910
Formula:
(CH2CH2NH)n Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00084427 Núm. CAS: 9002-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...