Zeolite ferrierite, ammonium
Proveedor: Thermo Fisher Scientific
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 400 m2/g, 20:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene
Formula:
SiO₂:Al₂O₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00132601 Núm. CAS: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...