Copper(II) ethoxide ≥98%
Proveedor: Thermo Fisher Scientific
|
Atención
|
acondicionado en Argón
46316.06
46316.14
46316.06EA
147
EUR
46316.06
46316.14
Copper(II) ethoxide ≥98%
Copper(II) ethoxide
Purity: 98%
CAS No.: 2850-65-9
Molecular Formula: C4H10CuO2
Formula Weight: 153.67
Physical Form: Powder, Packaged under Argon
Sensitivity: Air and Moisture Sensitive
Appearance: Green
Melting Point: ca 120° dec.
UN No.: UN3181
MDL No.: MFCD00015650
Formula:
C₄H₁₀CuO₂ Pto. de fusión: ∼ 120 °C (desc.) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00015650 Núm. CAS: 2850-65-9 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...