Polyester methacrylate PEM-665

Proveedor: Thermo Scientific
Atención

46408.22 46408.14
46408.22EA 805 EUR
46408.22 46408.14
Polyester methacrylate PEM-665
Polyester methacrylate

Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pale yellow


Applications: Good solubility in both aliphatic and aromatic co-monomers

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR