Polyester acrylate/methacrylate PEAM-645
Proveedor: Thermo Scientific
|
Atención
|
46596.22
46596.14
46596.22EA
324
EUR
46596.22
46596.14
Polyester acrylate/methacrylate PEAM-645
Polyester acrylate/methacrylate
CAS No.: 921213-39-0
Formula Weight: 645
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Dark amber to brown
Odour: Mild
Melting Point:
Applications: Recommended for use as a base resin in adhesive applications or coating applications
MW:
645 g/mol Pto. de fusión: >375 °C (desc.) Temperatura de almacenaje: Nevera |
Núm. CAS:
921213-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...