Zirconium(IV) isopropoxide isopropanol complex
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
Zirconium(IV) i-propoxide (isopropanol adduct)
86118.09
86118.18
86118.09EA
150
EUR
86118.09
86118.18
Zirconium(IV) isopropoxide isopropanol complex
Zirconium(IV) isopropoxide isopropanol complex
CAS No.: 14717-56-7
Molecular Formula: Zr[OCH(CH3)2]4.(CH3)2CHOH
Formula Weight: 387.67
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Boiling Point: dec.
Solubility: Soluble in hot Isopropanol, warm hexane and toluene. Tends to precipitate over time
UN No.: UN3181
MDL No.: MFCD00075157
Formula:
C₁₅H₃₆O₅Zr MW: 387,672 g/mol Pto. Ebullición: Dec. Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00075157 Núm. CAS: 14717-56-7 EINECS: 218-522-4 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...