L-serine methyl ester hydrochloride 98%

Proveedor: Thermo Scientific

Sinónimos: H-Ser-OMe.HCl

A12355.06 A12355.14 A12355.22
A12355.06EA 46.4 EUR
A12355.06 A12355.14 A12355.22
L-serine methyl ester hydrochloride 98%
L-serine methyl ester hydrochloride

Purity: 98% CAS No.: 5680-80-8 Molecular Formula: C4H9NO3.HCl Formula Weight: 155.58 Storage Temperature: -30°C to -10°C Melting Point: 155° dec. Beilstein Reference: 3559591 Optical Rotation: +5° (c=2 in methanol) MDL No.: MFCD00063680

Formula: C₄H₉NO₃
MW: 155,58 g/mol
Pto. de fusión: 155 °C (desc.)
Temperatura de almacenaje: Congelador
Número MDL: MFCD00063680
Núm. CAS: 5680-80-8
EINECS: 227-140-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR