5-Aminolevulinic acid hydrochloride 99%
Proveedor: Thermo Fisher Scientific
Sinónimos:
5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
A16942.04
A16942.MC
A16942.ME
A16942.04EA
242
EUR
A16942.04
A16942.MC
A16942.ME
5-Aminolevulinic acid hydrochloride 99%
5-Aminolevulinic acid hydrochloride
Purity: 99%
CAS No.: 5451-09-2
Molecular Formula: C5H9NO3.HCl
Formula Weight: 167.59
Storage Temperature: 2°C to 8°C
Melting Point: ca 156° dec.
Beilstein Reference: 3690651
MDL No.: MFCD00012869
Formula:
C₅H₁₀ClNO₃ Pto. de fusión: ∼ 156 °C (desc.) |
Número MDL:
MFCD00012869 Núm. CAS: 5451-09-2 EINECS: 226-679-5 Índice Merck: 14,446 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...