Aliquat® 336 TG
Proveedor: Thermo Scientific
|
Peligro
|
Sinónimos:
Methyltrialkylammonium chloride, Tricaprylmethylammonium chloride, Trioctylmethylammonium chloride, Methyltrioctylammonium chloride
CAS No.: 63393-96-4
Molecular Formula: [CH3(CH2)7]3NCH3Cl
Formula Weight: 404.17
Flash Point: 132°(269°F)
Physical Form: Viscous liquid
Density: 0.880
Refractive Index: 1.4465
Sensitivity: Hygroscopic
Appearance: Colourless to yellow
Odour: Mild
Boiling Point:
Formula:
CH₃(CH₂)₇]₃NCH₃Cl MW: 404,17 g/mol Pto. Ebullición: >240 °C Densidad: 0,880 Pto. de inflamación: 132 °C (269 °F) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011862 Núm. CAS: 63393-96-4 EINECS: 264-120-7 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...