Janus green B
Proveedor: Thermo Fisher Scientific
A17391.06
A17391.14
A17391.06EA
71
EUR
A17391.06
A17391.14
Janus green B
Janus green B
CAS No.: 2869-83-2
Molecular Formula: C30H31ClN6
Formula Weight: 511.07
MDL No.: MFCD00011758
Formula:
C₃₀H₃₁ClN₆ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00011758 Núm. CAS: 2869-83-2 EINECS: 220-695-6 Índice Merck: 14,5255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...