N-Carbobenzoxy-L-glutamine 99%
Proveedor: Thermo Fisher Scientific
A18240.06
A18240.14
A18240.06EA
26.6
EUR
A18240.06
A18240.14
N-Carbobenzoxy-L-glutamine 99%
N-Carbobenzoxy-L-glutamine
Purity: 99%
CAS No.: 2650-64-8
Molecular Formula: C13H16N2O5
Formula Weight: 280.28
Melting Point: 134-137°
Beilstein Reference: 2061271
Optical Rotation: -7° (c=2 in ethanol)
MDL No.: MFCD00008043
Formula:
C₁₃H₁₆N₂O₅ Pto. de fusión: 134…137 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00008043 Núm. CAS: 2650-64-8 EINECS: 220-173-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...