Inulin

Proveedor: Thermo Fisher Scientific

A18425.09 A18425.18 A18425.30
A18425.09EA 44.9 EUR
A18425.09 A18425.18 A18425.30
Inulin
Inulin

CAS No.: 9005-80-5 Physical Form: Powder Appearance: White Odour: Odourless MDL No.: MFCD00131407


Applications: Mixture of fructose polymers that serves as carbohydrate storage in plants

Temperatura de almacenaje: Ambiente
Número MDL: MFCD00131407
Núm. CAS: 9005-80-5
EINECS: 232-684-3
Índice Merck: 14,5004

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR