Formamidine disulphide dihydrochloride 97%
Proveedor: Thermo Scientific
|
Atención
|
Sinónimos:
Dithiourea Dihydrochloride, Dithiodiformamide Dihydrochloride
B20331.06
B20331.14
B20331.22
B20331.06EA
56.5
EUR
B20331.06
B20331.14
B20331.22
Formamidine disulphide dihydrochloride 97%
Formamidine disulphide dihydrochloride
Purity: 97%
CAS No.: 14807-75-1
Molecular Formula: C2H6N4S2.2HCl
Formula Weight: 223.15
Melting Point: ca 175° dec.
MDL No.: MFCD00012586
Formula:
C₂H₆N₄S₂ MW: 223,13 g/mol Pto. de fusión: ∼ 175 °C (desc.) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00012586 Núm. CAS: 14807-75-1 EINECS: 238-876-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...