Potassium-1-carboxyvinyl hydrogenphosphate 99%
Proveedor: Thermo Scientific
Sinónimos:
Phosphoenolpyruvate potassium salt, Potassium 1-carboxyvinyl hydrogenphosphate, Phosphoenolpyruvic acid monopotassium salt, Phosphoenolpyruvic acid potassium
Purity: 99%
CAS No.: 4265-07-0
Molecular Formula: C3H4KO6P
Formula Weight: 206.14
Storage Temperature: -30°C to -10°C
Melting Point: ca 170° dec.
Beilstein Reference: 4603446
MDL No.: MFCD00044476
Formula:
C₃H₄KO₆P MW: 206,131 g/mol Pto. de fusión: ∼ 170 °C (desc.) |
Número MDL:
MFCD00044476 Núm. CAS: 4265-07-0 EINECS: 224-247-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...