Ethylhydrocupreine hydrochloride 97%
Proveedor: Thermo Fisher Scientific
B21627.MC
B21627.ME
B21627.MCEA
32.3
EUR
B21627.MC
B21627.ME
Ethylhydrocupreine hydrochloride 97%
Ethylhydrocupreine hydrochloride
Purity: 97%
CAS No.: 3413-58-9
Molecular Formula: C21H26N2O2.HCl
Formula Weight: 374.91
Storage Temperature: 2°C to 8°C
Melting Point: ca 240° dec.
MDL No.: MFCD00135594
Formula:
C₂₁H₂₆N₂O₂·HCl Pto. de fusión: ∼ 240 °C (desc.) |
Número MDL:
MFCD00135594 Núm. CAS: 3413-58-9 EINECS: 222-302-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...