1-Adamantylacetonitrile 97%
Proveedor: Thermo Fisher Scientific
|
Peligro
|
B24770.03
B24770.06
B24770.03EA
86.2
EUR
B24770.03
B24770.06
1-Adamantylacetonitrile 97%
1-Adamantylacetonitrile
Purity: 97%
CAS No.: 16269-13-9
Molecular Formula: C12H17N
Formula Weight: 175.28
UN No.: UN3439
MDL No.: MFCD00194034
Formula:
C₁₂H₁₇N Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00194034 Núm. CAS: 16269-13-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...