1-Benzyl-N-methyl-4-piperidinamine 98%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H27606.06
H27606.03
H27606.06EA
97.1
EUR
H27606.06
H27606.03
1-Benzyl-N-methyl-4-piperidinamine 98%
1-Benzyl-N-methyl-4-piperidinamine
Purity: 98%
CAS No.: 7006-50-0
Molecular Formula: C13H20N2
Formula Weight: 204.31
MDL No.: MFCD03931054
Formula:
C₁₃H₂₀N₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03931054 Núm. CAS: 7006-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...