(R)-(+)-2-Methyl-2-propanesulphinamide 98%
Proveedor: Thermo Scientific
Sinónimos:
(R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide
H27724.03
H27724.06
H27724.14
H27724.03EA
73.7
EUR
H27724.03
H27724.06
H27724.14
(R)-(+)-2-Methyl-2-propanesulphinamide 98%
(R)-(+)-2-Methyl-2-propanesulphinamide
Purity: 98%
CAS No.: 196929-78-9
Molecular Formula: C4H11NOS
Formula Weight: 121.20
Storage Temperature: 2°C to 8°C
Melting Point: ca 100-105°
MDL No.: MFCD05861479
Formula:
C₄H₁₁NOS MW: 121,20 g/mol Pto. de fusión: ∼ 100…105 °C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD05861479 Núm. CAS: 196929-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...