3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde 95%
Proveedor: Thermo Fisher Scientific
H32167.03
H32167.06
H32167.03EA
107
EUR
H32167.03
H32167.06
3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde 95%
3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde
Purity: 95%
CAS No.: 67869-90-3
Molecular Formula: C10H10O3
Formula Weight: 178.19
Boiling Point: 120-122°
MDL No.: MFCD01893356
Formula:
C₁₀H₁₀O₃ Pto. Ebullición: 120…122 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01893356 Núm. CAS: 67869-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...