Glycine dimethylamide 97%

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: 2-Amino-N,N-dimethylacetamide

H50293.03 H50293.MD
H50293.03EA 173 EUR
H50293.03 H50293.MD
Glycine dimethylamide 97%
Glycine dimethylamide

Purity: 97% CAS No.: 1857-19-8 Molecular Formula: C4H10N2O Formula Weight: 102.14 Odour: Amine-like UN No.: UN2735 MDL No.: MFCD06655265

Formula: C₄H₁₀N₂O
MW: 102.14 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06655265
Núm. CAS: 1857-19-8
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR